Learn More
Medchemexpress LLC Butaxamine hydrochloride | 5696-15-1 | 99.9% | 303.82 g/mol | C15H26ClNO3 | 5 MG
SDP
Catalog No. 5000308310
Encompass_Preferred
Butaxamine hydrochloride is the hydrochloride salt of butaxamine, a selective β2-adrenergic receptor antagonist used in pharmacological research to probe β2-adrenergic signaling and physiological responses. It is provided as a high-purity solid suitable for small-scale laboratory experiments.
  • Selective β2-adrenergic receptor antagonist.
  • Intended for in vitro and in vivo pharmacology studies.
  • High purity (99.87%) for reproducible results.
  • Supplied in small 5 mg quantities for bench-scale use.
  • White to off-white solid, hydrochloride salt improves solubility.
  • Store at 4°C, sealed, and protect from moisture.
Catalog No. 50-003-08310 Supplier Medchemexpress LLC Supplier No. HY1184705MG
Butaxamine hydrochloride is the hydrochloride salt of butaxamine, a selective β2-adrenergic receptor antagonist used in pharmacological research to probe β2-adrenergic signaling and physiological responses. It is provided as a high-purity solid suitable for small-scale laboratory experiments.
  • Selective β2-adrenergic receptor antagonist.
  • Intended for in vitro and in vivo pharmacology studies.
  • High purity (99.87%) for reproducible results.
  • Supplied in small 5 mg quantities for bench-scale use.
  • White to off-white solid, hydrochloride salt improves solubility.
  • Store at 4°C, sealed, and protect from moisture.