Learn More
Medchemexpress LLC HY-16558 5mg Medchemexpress, Butein CAS:487-52-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY165585MG
Encompass_Preferred
Medchemexpress, HY-16558 5mg Butein CAS:487-52-5 Butein, isolated from Dalbergia odorifera T. Chen, is a cAMP-specific PDE inhibitor with an IC50 of 10.4 μM for PDE4. Butein is a specific protein tyrosine kinase inhibitor with IC50s of 16 and 65 μM for EGFR and p60c-src in HepG2 cells. Butein sensitizes HeLa cells to Cisplatin through AKT and ERK/p38 MAPK pathways by targeting FoxO3a. Butein is a SIRT1 activator (STAC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1892