Learn More
Apexbio Technology LLC Butenafine HCl 101827-46-7 5g
SDP
Catalog No. 5000094298
Encompass_Preferred
Butenafine HCl (CAS 101827-46-7) is a small-molecule inhibitor targeting squalene epoxidase It is designed to inhibit this enzyme thereby disrupting fungal sterol biosynthesis Butenafine HCl exerts its biological activity primarily through inhibition of squalene epoxidase leading to impaired ergosterol synthesis and accumulation of squalene which disrupts fungal cell membrane integrity and results in cell death In biochemical studies Butenafine HCl demonstrates inhibitory activity with reported IC50 values ranging from approximately 0 03 M to 0 1 M depending on fungal species and experimental conditions Based on these pharmacological properties Butenafine HCl holds research potential in biomedical antifungal studies including investigations into fungal physiology antifungal efficacy and drug resistance mechanisms
Catalog No. 50-000-94298 Supplier Apexbio Technology LLC Supplier No. B19015000
Butenafine HCl (CAS 101827-46-7) is a small-molecule inhibitor targeting squalene epoxidase It is designed to inhibit this enzyme thereby disrupting fungal sterol biosynthesis Butenafine HCl exerts its biological activity primarily through inhibition of squalene epoxidase leading to impaired ergosterol synthesis and accumulation of squalene which disrupts fungal cell membrane integrity and results in cell death In biochemical studies Butenafine HCl demonstrates inhibitory activity with reported IC50 values ranging from approximately 0 03 M to 0 1 M depending on fungal species and experimental conditions Based on these pharmacological properties Butenafine HCl holds research potential in biomedical antifungal studies including investigations into fungal physiology antifungal efficacy and drug resistance mechanisms