Learn More
Medchemexpress LLC Butenafine (hydrochloride) | 101827-46-7 | 100.0% | 353.93 | 1 ML
SDP
Catalog No. 5000385956
Encompass_Preferred
Butenafine Hydrochloride is a synthetic benzylamine antifungal. It works by inhibiting the synthesis of sterols through the inhibition of squalene epoxidase. This drug is used to control dermal fungal infections such as athlete's foot and ringworm. It has excellent penetration into the epidermis and a prolonged retention time, conferring residual therapeutic activity after treatment cessation. Butenafine also possesses anti-inflammatory activity.
  • Inhibits squalene epoxidase
  • Controls dermal fungal infections
  • Provides excellent epidermal penetration
  • Offers prolonged retention time
  • Possesses anti-inflammatory properties
  • Effective for tinea corporis cruris and tinea manuum pedis
Catalog No. 50-003-85956 Supplier Medchemexpress LLC Supplier No. HY1739610MM/1ML
Butenafine Hydrochloride is a synthetic benzylamine antifungal. It works by inhibiting the synthesis of sterols through the inhibition of squalene epoxidase. This drug is used to control dermal fungal infections such as athlete's foot and ringworm. It has excellent penetration into the epidermis and a prolonged retention time, conferring residual therapeutic activity after treatment cessation. Butenafine also possesses anti-inflammatory activity.
  • Inhibits squalene epoxidase
  • Controls dermal fungal infections
  • Provides excellent epidermal penetration
  • Offers prolonged retention time
  • Possesses anti-inflammatory properties
  • Effective for tinea corporis cruris and tinea manuum pedis