Learn More
Medchemexpress LLC Butenafine (Hydrochloride) | 101827-46-7 | 353.93 | 500 MG
SDP
Catalog No. 5000365685
Encompass_Preferred
Butenafine Hydrochloride is a synthetic benzylamine antifungal that works by inhibiting squalene epoxidase, thereby inhibiting the synthesis of sterols. It is used to control dermal fungal infections like athlete's foot and ringworm, and also possesses anti-inflammatory activity. The drug has excellent penetration into the epidermis and prolonged retention time after topical application, conferring residual therapeutic activity.
  • Inhibits sterol synthesis by inhibiting squalene epoxidase
  • Targets fungal infections such as athlete's foot and ringworm
  • Provides excellent penetration into the epidermis
  • Offers prolonged retention time after topical application
  • Possesses anti-inflammatory activity
Catalog No. 50-003-65685 Supplier Medchemexpress LLC Supplier No. 500MGHY17396
Butenafine Hydrochloride is a synthetic benzylamine antifungal that works by inhibiting squalene epoxidase, thereby inhibiting the synthesis of sterols. It is used to control dermal fungal infections like athlete's foot and ringworm, and also possesses anti-inflammatory activity. The drug has excellent penetration into the epidermis and prolonged retention time after topical application, conferring residual therapeutic activity.
  • Inhibits sterol synthesis by inhibiting squalene epoxidase
  • Targets fungal infections such as athlete's foot and ringworm
  • Provides excellent penetration into the epidermis
  • Offers prolonged retention time after topical application
  • Possesses anti-inflammatory activity