Learn More
Medchemexpress LLC Butenafine Hydrochloride | 101827-46-7 | 99.9% | 353.93 | 1 G
SDP
Catalog No. 5000384056
Encompass_Preferred
Butenafine Hydrochloride is a synthetic benzylamine antifungal that inhibits the synthesis of sterols by inhibiting squalene epoxidase. It is effective against dermal fungal infections such as athlete's foot and ringworm. The drug offers excellent epidermal penetration and prolonged retention, providing residual therapeutic activity. It also possesses anti-inflammatory activity.
  • Inhibits sterol synthesis
  • Effective against dermal fungal infections
  • Excellent epidermal penetration
  • Prolonged retention time
  • Possesses anti-inflammatory activity
  • Safe and effective for tinea corporis cruris and tinea manuum pedis
Catalog No. 50-003-84056 Supplier Medchemexpress LLC Supplier No. HY173961G
Butenafine Hydrochloride is a synthetic benzylamine antifungal that inhibits the synthesis of sterols by inhibiting squalene epoxidase. It is effective against dermal fungal infections such as athlete's foot and ringworm. The drug offers excellent epidermal penetration and prolonged retention, providing residual therapeutic activity. It also possesses anti-inflammatory activity.
  • Inhibits sterol synthesis
  • Effective against dermal fungal infections
  • Excellent epidermal penetration
  • Prolonged retention time
  • Possesses anti-inflammatory activity
  • Safe and effective for tinea corporis cruris and tinea manuum pedis