Learn More
Cayman Chemical Butenoyl PAF, Purity: ≥98%, CAS: 474944-25-7, 50 mg
SDP
Catalog No. 502893699
Encompass_Preferred
A product of the oxidative decomposition of 2-arachidonoyl phospholipids; 10-fold less potent than PAF as a PAF receptor agonist, but present in amounts 100-fold greater
Catalog No. 50-289-3699 Supplier Cayman Chemical Supplier No. 6092950 MG
A product of the oxidative decomposition of 2-arachidonoyl phospholipids; 10-fold less potent than PAF as a PAF receptor agonist, but present in amounts 100-fold greater