Learn More
Enzo Life Sciences Buthionine sulfoximine (500mg). CAS: 83730-53-4
SDP
Supplier:  Enzo Life Sciences BMLFR1170500
Encompass_Preferred
L-Buthionine-sulfoximine (BSO) induces oxidative stress in cells by irreversibly inhibiting γ-glutamylcysteine synthetase, an essential enzyme for the synthesis of glutathione. Alternative name: BSO. Purity: ≥97% (TLC). Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-0838