Learn More
Cayman Chemical L-Buthionine-(S,R)-Sulfoximine, 83730-53-4, 500 mg
SDP
Supplier:  Cayman Chemical 14484500
Encompass_Preferred
Molecular Formula C8H18N2O3S, Purity ≥98%, Formula Weight 222.3, Synonyms: NSC 326231; An irreversible inhibitor of -glutamylcysteine synthetase (Ki <100 M), the rate-limiting enzyme for GSH synthesis, that induces oxidative stress in cells by depleting GSH; enhances the toxicity of various chemotherapeutic agents in drug-resistant tumors.
Catalog No. NC0859739