Learn More
Medchemexpress LLC Butin | 21913-99-5 | 5 MG
SDP
Catalog No. 5000418876
Encompass_Preferred
Butin is a major biologically active flavonoid isolated from the heartwood of *Dalbergia odorifera*. It exhibits oral activity and possesses strong antioxidant, antiplatelet, and anti-inflammatory properties. It has been shown to significantly alleviate myocardial infarction, improve heart function, and prevent diabetes-induced cardiac oxidative damage in rats.
  • Isolated from the heartwood of *Dalbergia odorifera*.
  • Exhibits strong antioxidant activity.
  • Possesses antiplatelet activity.
  • Has anti-inflammatory activity.
  • Alleviates myocardial infarction.
  • Improves heart function.
  • Prevents diabetes-induced cardiac oxidative damage in rats.
  • Reduces cell apoptosis and death caused by ischemia/reperfusion injury.
  • Disrupts interaction between Nrf2 and Keap1.
  • Increases expression of antioxidant genes.
  • Purity: 99.34%.
Catalog No. 50-004-18876 Supplier Medchemexpress LLC Supplier No. HYN6020B5MG
Butin is a major biologically active flavonoid isolated from the heartwood of *Dalbergia odorifera*. It exhibits oral activity and possesses strong antioxidant, antiplatelet, and anti-inflammatory properties. It has been shown to significantly alleviate myocardial infarction, improve heart function, and prevent diabetes-induced cardiac oxidative damage in rats.
  • Isolated from the heartwood of *Dalbergia odorifera*.
  • Exhibits strong antioxidant activity.
  • Possesses antiplatelet activity.
  • Has anti-inflammatory activity.
  • Alleviates myocardial infarction.
  • Improves heart function.
  • Prevents diabetes-induced cardiac oxidative damage in rats.
  • Reduces cell apoptosis and death caused by ischemia/reperfusion injury.
  • Disrupts interaction between Nrf2 and Keap1.
  • Increases expression of antioxidant genes.
  • Purity: 99.34%.