Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (+)-Butin | 74628-42-5 | 5 MG
SDP

Catalog No. 5000418584
Encompass_Preferred

(+)-Butin is the R-enantiomer of Butin (HY-N6020B). Butin is a bioactive flavonoid. Butin can be isolated from the heartwood of Dalbergia cocos. Butin exhibits potent antioxidant, antiplatelet, and anti-inflammatory activities.

  • R-enantiomer of butin
  • Bioactive flavonoid
  • Isolated from Dalbergia cocos
  • Exhibits potent antioxidant activity
  • Provides antiplatelet activity
  • Demonstrates anti-inflammatory activities
  • Purity of 99.54%
  • Enantiomeric excess of 98.30%
  • Molecular weight of 272.26
  • Molecular formula C15H12O5
  • Appears as a yellow to brown solid

Catalog No. 50-004-18584 Supplier Medchemexpress LLC Supplier No. HYN6020A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

(+)-Butin is the R-enantiomer of Butin (HY-N6020B). Butin is a bioactive flavonoid. Butin can be isolated from the heartwood of Dalbergia cocos. Butin exhibits potent antioxidant, antiplatelet, and anti-inflammatory activities.

  • R-enantiomer of butin
  • Bioactive flavonoid
  • Isolated from Dalbergia cocos
  • Exhibits potent antioxidant activity
  • Provides antiplatelet activity
  • Demonstrates anti-inflammatory activities
  • Purity of 99.54%
  • Enantiomeric excess of 98.30%
  • Molecular weight of 272.26
  • Molecular formula C15H12O5
  • Appears as a yellow to brown solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.