Learn More
Medchemexpress LLC (+)-Butin (Standard) | 74628-42-5 | 1 MG
SDP
Catalog No. 5000419910
Encompass_Preferred
(+)-Butin is the R-enantiomer of Butin, a bioactive flavonoid isolated from the heartwood of *Dalbergia cocos*. It demonstrates potent antioxidant, antiplatelet, and anti-inflammatory activities.
  • R-enantiomer of Butin
  • Bioactive flavonoid
  • Isolated from *Dalbergia cocos*
  • Exhibits antioxidant activity
  • Exhibits antiplatelet activity
  • Exhibits anti-inflammatory activity
  • For research use only
Catalog No. 50-004-19910 Supplier Medchemexpress LLC Supplier No. HYN6020A1MG
(+)-Butin is the R-enantiomer of Butin, a bioactive flavonoid isolated from the heartwood of *Dalbergia cocos*. It demonstrates potent antioxidant, antiplatelet, and anti-inflammatory activities.
  • R-enantiomer of Butin
  • Bioactive flavonoid
  • Isolated from *Dalbergia cocos*
  • Exhibits antioxidant activity
  • Exhibits antiplatelet activity
  • Exhibits anti-inflammatory activity
  • For research use only