Learn More
Electron Microscopy Sciences Butler Block Trimmer
SDP
Catalog No. 5024829
Encompass_Preferred
A precision made device to aid in hand trimming resin tissue blocks to the desired shape for ultramicrotomy. The knurled knife guide swivels to help the trimming blade to cut the sides of the block. May be utilized with any stereomicroscope with epi-illumination. The trimmer is supplied with a standard chuck to accommodate the block size 00. A flat embedding mold adapter is available as an option. (Stain Technology, Vol. 49, No 4: pp. 129-132, 1974)
Catalog No. 50-248-29 Supplier Electron Microscopy Sciences Supplier No. 6994501
A precision made device to aid in hand trimming resin tissue blocks to the desired shape for ultramicrotomy. The knurled knife guide swivels to help the trimming blade to cut the sides of the block. May be utilized with any stereomicroscope with epi-illumination. The trimmer is supplied with a standard chuck to accommodate the block size 00. A flat embedding mold adapter is available as an option. (Stain Technology, Vol. 49, No 4: pp. 129-132, 1974)