Learn More
Medchemexpress LLC HY-B0293 100mg Medchemexpress, Butoconazole (nitrate) CAS:64872-77-1 Purity:>98%
SDP
Catalog No. 502082452
Encompass_Preferred
Medchemexpress, HY-B0293 100mg Butoconazole (nitrate) CAS:64872-77-1 Butoconazole nitrate (RS 35887), an imidazole antifungal agent, is active against Candida spp. and effective against vaginal infections due to Candida albicans. Butoconazole nitrate is presumed to function as other imidazole derivatives via inhibition of steroid synthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2452 Supplier Medchemexpress LLC Supplier No. HYB0293100MG
Medchemexpress, HY-B0293 100mg Butoconazole (nitrate) CAS:64872-77-1 Butoconazole nitrate (RS 35887), an imidazole antifungal agent, is active against Candida spp. and effective against vaginal infections due to Candida albicans. Butoconazole nitrate is presumed to function as other imidazole derivatives via inhibition of steroid synthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.