Learn More
TARGETMOL CHEMICALS INC Butoconazole nitrate 500MG
SDP
Catalog No. 502878745
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Butoconazole Nitrate is the nitrate salt form of butoconazole, a synthetic imidazole derivative with fungistatic properties. Butoconazole nitrate (RS 35887) interferes with steroid biosynthesis by inhibiting the conversion of lanosterol to ergosterol, thereby changing the fungal cell membrane lipid composition. This alters cell permeability and leads to growth inhibition. Butoconazole nitrate is active against many dermatophytes and yeasts. It also contains antibacterial effects against some gram-positive organisms. Purity 100%
Catalog No. 50-287-8745 Supplier TARGETMOL CHEMICALS INC Supplier No. T1438500MG
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Butoconazole Nitrate is the nitrate salt form of butoconazole, a synthetic imidazole derivative with fungistatic properties. Butoconazole nitrate (RS 35887) interferes with steroid biosynthesis by inhibiting the conversion of lanosterol to ergosterol, thereby changing the fungal cell membrane lipid composition. This alters cell permeability and leads to growth inhibition. Butoconazole nitrate is active against many dermatophytes and yeasts. It also contains antibacterial effects against some gram-positive organisms. Purity 100%