Learn More
Medchemexpress LLC Butoconazole nitrate | 64872-77-1 | 99.4% | 200 MG
SDP
Catalog No. 5000408993
Encompass_Preferred
Butoconazole nitrate is an imidazole antifungal agent that is active against Candida spp., including *Candida albicans*. It functions by inhibiting steroid synthesis, specifically the conversion of lanosterol to ergosterol, which alters fungal cell membrane lipid composition, leading to changes in cell permeability and ultimately growth inhibition of the fungal cell.
  • Active against *Candida* species
  • Effective for vaginal infections caused by *Candida albicans*
  • Inhibits fungal steroid synthesis
  • Alters fungal cell membrane lipid composition
Catalog No. 50-004-08993 Supplier Medchemexpress LLC Supplier No. HYB0293200MG
Butoconazole nitrate is an imidazole antifungal agent that is active against Candida spp., including *Candida albicans*. It functions by inhibiting steroid synthesis, specifically the conversion of lanosterol to ergosterol, which alters fungal cell membrane lipid composition, leading to changes in cell permeability and ultimately growth inhibition of the fungal cell.
  • Active against *Candida* species
  • Effective for vaginal infections caused by *Candida albicans*
  • Inhibits fungal steroid synthesis
  • Alters fungal cell membrane lipid composition