Learn More
Medchemexpress LLC Butoconazole nitrate | 64872-77-1 | MFCD00058159 | 99.87% | 1 ML
SDP
Catalog No. 5000406481
Encompass_Preferred
Butoconazole nitrate is an imidazole antifungal agent that is active against Candida spp. and effective against vaginal infections caused by Candida albicans. It functions by inhibiting steroid synthesis, similar to other imidazole derivatives. This action changes the fungal cell membrane lipid composition, which in turn alters cell permeability and leads to osmotic disruption or growth inhibition of the fungal cell.
  • Imidazole antifungal agent
  • Active against Candida spp.
  • Effective against vaginal infections caused by Candida albicans
  • Inhibits steroid synthesis
  • Alters fungal cell membrane lipid composition
  • Leads to osmotic disruption or growth inhibition of fungal cells
Catalog No. 50-004-06481 Supplier Medchemexpress LLC Supplier No. HYB029310MM/1ML
Butoconazole nitrate is an imidazole antifungal agent that is active against Candida spp. and effective against vaginal infections caused by Candida albicans. It functions by inhibiting steroid synthesis, similar to other imidazole derivatives. This action changes the fungal cell membrane lipid composition, which in turn alters cell permeability and leads to osmotic disruption or growth inhibition of the fungal cell.
  • Imidazole antifungal agent
  • Active against Candida spp.
  • Effective against vaginal infections caused by Candida albicans
  • Inhibits steroid synthesis
  • Alters fungal cell membrane lipid composition
  • Leads to osmotic disruption or growth inhibition of fungal cells