Learn More
MAPA Professional Butoflex 651 Gloves
Catalog No. 17111161 Shop All MAPA Professional Products
Change view
Click to view available options
Size:
7
Catalog No. Size
17-111-161 7
1 options
Chemical Resistant
Catalog No. 17-111-161 Supplier MAPA Professional Supplier No. 5630
Add to Cart
Add to Cart
Chemical Resistant
Multi-layered black butyl glove that is unsupported with Z-grip
  • Formulating paints, varnishes, resins and inks
  • Handling and repackaging chemicals
  • Sampling chemicals
Specifications
Cleanroom Class Class 1 (ISO 3)
Accelerator Free Yes
AQL None
Color Black
Disposable Reusable
Exterior Finish Smooth
Finger Thickness (English) 22 mil
Finger Thickness (Metric) 0.56 mm
Hand Ambidextrous
Latex Free Yes
Material Co-polymer
Palm Thickness (English) 22 mil
Palm Thickness (Metric) 0.56 mm
Powdered Powder Free
Product Type Controlled Environment Gloves
Size 7
Sterility Non-sterile
Thickness (English) 22 mil
Thickness (Metric) 0.56 mm