Learn More
Sigma Aldrich Fine Chemicals Biosciences ButojectTM butyl gloves si
Catalog No. 502901655
Encompass
Butoject is a highly flexible and effective chemical protective glove made of Butyl rubber suitable for extreme demands for work with esters and ketones. With a thickness of merely 0.30 mm and very good cold flexibility Butoject is very comfortable to wear and allows good dexterity. To improve the grip on smooth and wet parts this glove has an additionally roughened surface.Very good protection against extremely aggressive chemicalsErgonomic fit and high elasticity ensures excellent wearing comfortRoughened surface for firm grip when wetGood mechanical resistanceCE compliant
Catalog No. 50-290-1655 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6776471PR
Butoject is a highly flexible and effective chemical protective glove made of Butyl rubber suitable for extreme demands for work with esters and ketones. With a thickness of merely 0.30 mm and very good cold flexibility Butoject is very comfortable to wear and allows good dexterity. To improve the grip on smooth and wet parts this glove has an additionally roughened surface.Very good protection against extremely aggressive chemicalsErgonomic fit and high elasticity ensures excellent wearing comfortRoughened surface for firm grip when wetGood mechanical resistanceCE compliant