Learn More
Electron Microscopy Sciences 2-Butoxy Ethanol 450 ML
SDP
Catalog No. 50980266
Encompass_Preferred
(Butyl Cellosolve; Ethylene Glycol Monobutyl Ether)
CH₃(CH₂)₃OCH₂CH₂OH F.W. 118.18
Boiling point: 169°C
Also known as Butyl Cellosolve and Ethylene Glycol Monobutyl Ether.
Catalog No. 50-980-266 Supplier Electron Microscopy Sciences Supplier No. 11820
(Butyl Cellosolve; Ethylene Glycol Monobutyl Ether)
CH₃(CH₂)₃OCH₂CH₂OH F.W. 118.18
Boiling point: 169°C
Also known as Butyl Cellosolve and Ethylene Glycol Monobutyl Ether.