Learn More
AdipoGen Butroxydim
SDP
Supplier:  AdipoGen CDXB0126M100
Encompass_Preferred
Chemical. CAS 138164-12-2. Formula C24H33NO4. MW 399.5. Synthetic. Cyclohexene oxime herbicide. Inhibitor of acetyl CoA carboxylase ACCase, a key enzyme in the synthesis of fatty acids. Compound can be used as analytical reference material.
Catalog No. 50-256-1664