Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butter Yellow, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018939 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8939 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8939 Supplier MilliporeSigma Supelco Supplier No. 7322525MG
Add to Cart
Add to Cart

Butter Yellow is a food dye widely used in industrial food production and is found to be carcinogenic in nature.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 60-11-7
Physical Form Neat
Linear Formula C6H5 N=NC6H4 N(CH3)2
MDL Number MFCD00008308
Melting Point 111°C
Synonym Dimethyl yellow; 4-(Dimethylamino)azobenzene; N,N-Dimethyl-4-(phenylazo)aniline; Butter yellow; Methyl yellow
Type Analytical Standard
Quantity 25 mg
Formula Weight 225.29
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.