Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Butterflies in the Classroom Kit Refill
SDPSCI_ED

Catalog No. S28651 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S28651 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S28651 Supplier Carolina™ Supplier No. 144071

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Replenishes the Butterflies in the Classroom kits.

Components:
  • Painted Lady Butterfly Culture (33 larvae with food) : 1
  • Painted Lady Butterfly Culture Medium (food): 1
  • Cups with Lids, 28.3g: 33
  • Wooden Craft Sticks : 2
  • Sorting Brushed: 2
  • Kimwipes™: 10
  • Each kit includes 1 year of FREE single-teacher license access to butterfly videos and images that can be used with interactive whiteboards and teacher presentation stations

Specifications

Quantity 1 kit
Includes 33 to 35 larvae, culture medium, 33 culture vessels with lids, filter paper, and 2 spatulas
Sufficient For Class of 30 students
Product Type Butterflies in the Classroom Kit (with Live Caterpillars)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.