Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roylco™ Butterfly Ornaments Craft Kit
SDPSCI_ED

Catalog No. S13227
Change view
Click to view available options
Grade:
1+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S13227 1+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13227 Supplier Roylco™ Supplier No. R83260
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A complete butterfly craft kit that features Bug Bodies, pre-cut Color Diffusing Paper™ wings, and antennae!

  • Color the wings with watercolor paint or washable markers, then spray with water
  • Watch the colors blend into unique designs
  • Once the wings are dry, fold and thread them through the slot in the bug body
  • Fluff out the wings on either side
  • For the finishing touch, thread the antenna through the head
  • Attach a chenille stem through the body to turn the bug into a beautiful bracelet or hair bow
  • Dimensions of Bug Bodies: 0.5 x 2 in. (1 x 5 cm)

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade 1+
Format Kit
Includes 75 Bug Bodies, Color Diffusing Wings, Antennae, Guide
Quantity 75 Butterflies
Languages English, Spanish, French, German
Subject Biology - Insects
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.