Learn More
Electron Microscopy Sciences Butvar-B-98 Powder 100 GR
SDP
Catalog No. 50980267
Encompass_Preferred
Polyvinyl butyl resin for thin film support. Stronger than Formvar and more sticky, resulting in a firm attachment for slotted or meshed grids. The standard solution is 0.25% Butvar B-98 in Chloroform. Handley, D.A., et. al., Ultramicroscopy, 4,479 (1979).
Catalog No. 50-980-267 Supplier Electron Microscopy Sciences Supplier No. 11850
Polyvinyl butyl resin for thin film support. Stronger than Formvar and more sticky, resulting in a firm attachment for slotted or meshed grids. The standard solution is 0.25% Butvar B-98 in Chloroform. Handley, D.A., et. al., Ultramicroscopy, 4,479 (1979).