Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc BUTYL 15UM 200A 500X30MM
SDP

Catalog No. 5090005097
Encompass

Butyl 15Um; 200A 500 X 30mm

Catalog No. 50-900-05097 Supplier YMC America, Inc Supplier No. BU20S165030WT
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Butyl 15Um; 200A 500 X 30mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.