Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl 2-Methylvalerate 98.0+%, TCI America™
SDP

Catalog No. B3359500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3359500ML 500 mL
B335925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3359500ML Supplier TCI America Supplier No. B3359500ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6297-41-2
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.268
InChI Key FMVUGLMJUHFPSJ-UHFFFAOYSA-N
Synonym 2-Methylvaleric Acid Butyl Ester, Butyl 2-Methylpentanoate, 2-Methylpentanoic Acid Butyl Ester
PubChem CID 95539
IUPAC Name butyl 2-methylpentanoate
SMILES CCCCOC(=O)C(C)CCC

Specifications

Color Colorless
Quantity 500 mL
Formula Weight 172.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl 2-Methylvalerate
EINECSNumber (2)-0782
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.