Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Butyl 3-mercaptopropionate

Catalog No. 381454500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-145-4500ML 500 mL
38-145-4100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 38-145-4500ML Supplier Sigma Aldrich Supplier No. 381454500ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Butyl 3-mercaptopropionate (3MPA) is a monofunctional thiol that can be used as a cross-linker and chain transferring agent for controlling the molecular weight of the polymer. It can be used in the thiolene photopolymerization.3MPA can be used as a ligand which can functionalize the quantum dots for the development of high luminescence light emitting diodes. It can also be used as a crosslinking monomeric unit for the preparation of thiol-acrylate based photopolymers.

Specifications

CAS 16215-21-7
Density 0.999 g/mL (at 25°C)
Boiling Point 101°C (2 mmHg)
Molecular Formula C7H14O2S
Refractive Index n20/D 1.457 (literature)
Linear Formula HSCH2CH2CO2(CH2)3 CH3
Quantity 500 mL
MDL Number MFCD00083561
Synonym Butyl 3-mercaptopropanoate
Molecular Weight (g/mol) 162.25
Percent Purity 98%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.