Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc BUTYL 3UM 120A 100X2MM
SDP

Catalog No. 50874344
Encompass_Preferred

Butyl 3Um; 120A; 100 X 2mm

Catalog No. 50-874-344 Supplier YMC America, Inc Supplier No. BU12S031002WT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Butyl 3Um; 120A; 100 X 2mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.