Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc BUTYL 3UM 120A 50X3MM
SDP

Catalog No. 50874343
Encompass_Preferred

Butyl 3Um; 120A; 50 X 3mm

Catalog No. 50-874-343 Supplier YMC America, Inc Supplier No. BU12S030503WT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Butyl 3Um; 120A; 50 X 3mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.