Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Butyl 4-hydroxybenzoate

Catalog No. 54680250GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-680-250GF 250 g
54-680-50GF 50 g
54-680-1KGF 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 54-680-250GF Supplier Sigma Aldrich Supplier No. 54680250GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Butyl 4-hydroxybenzoate

Specifications

CAS 94-26-8
Melting Point 67°C to 70°C (lit.), 68°C to 70°C
Boiling Point 156°C to 157°C (3.5 mmHg)
Molecular Formula C11H14O3
Linear Formula HOC6H4CO2(CH2)3 CH3
MDL Number MFCD00016478
Quantity 250 g
Synonym p-Hydroxybenzoic acid n-butyl ester; Butyl paraben; SPF
Molecular Weight (g/mol) 194.23
Percent Purity ≥99.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.