Learn More
Cayman Chemical Butyl 4-hydroxybenzoate, Purity: ≥98%, CAS: 94-26-8, 250 g
SDP
Catalog No. 502909682
Encompass_Preferred
A paraben; active against B. subtilis, S. aureus, C. molischiana, S. cerevisiae, A. niger, P. chrysogenum (MICs = 75, 75, 60, 52, 55, and 45 mg/L, respectively); decreases the viability of, and ATP and GSH levels in, HepG2 and HDFn cells at 300-1,000 µM; binds to estrogen receptors; increases uterine weight in immature and ovariectomized rats; has been found in influent and effluent wastewater
Catalog No. 50-290-9682 Supplier Cayman Chemical Supplier No. 36797250 G
A paraben; active against B. subtilis, S. aureus, C. molischiana, S. cerevisiae, A. niger, P. chrysogenum (MICs = 75, 75, 60, 52, 55, and 45 mg/L, respectively); decreases the viability of, and ATP and GSH levels in, HepG2 and HDFn cells at 300-1,000 µM; binds to estrogen receptors; increases uterine weight in immature and ovariectomized rats; has been found in influent and effluent wastewater