Learn More
Butyl 4-hydroxybenzoate-ring-13C6 Solution, 50 μg/mL in Acetone, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| CAS | 1416711-53-9 |
| Flash Point | -17.0°C - Closed Cup |
| Linear Formula | 13C6C5H14O3 |
| Synonym | 4-Hydroxybenzoic acid n-butyl ester-ring-13C6 solution; Butyl paraben-ring-13C6 solution |
| Type | Analytical Standard |
| Quantity | 1 mL |
| Formula Weight | 200.18 |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 50 μg/mL in acetone |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.