Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc BUTYL 5UM 300A 30X10MM
SDP

Catalog No. 5090005111
Encompass_Preferred

Butyl 5Um; 300A 30 X 10mm

Catalog No. 50-900-05111 Supplier YMC America, Inc Supplier No. BU30S050310WTG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Butyl 5Um; 300A 30 X 10mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.