Learn More
Sigma Aldrich Fine Chemicals Biosciences Butyl acetate anhydrous 99
Catalog No. 502899048
Encompass
Butyl acetate is an aliphatic linear ester commonly used as an organic solvent in cosmetic and pharmaceutical industries and as a flavoring agent in food industry. It is a promising candidate as biofuel additive.
Catalog No. 50-289-9048 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2877252L
Butyl acetate is an aliphatic linear ester commonly used as an organic solvent in cosmetic and pharmaceutical industries and as a flavoring agent in food industry. It is a promising candidate as biofuel additive.