Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl acetate, CHROMASOLV™ Plus, for HPLC, 99.7%, Solstice

Catalog No. 6001012 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-010-12 6 x 1 L Glass Bottle
60-010-11 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-010-12 Supplier Solstice Supplier No. 2706876X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHROMASOLV™ Plus, for HPLC, 99.7%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009445
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
PubChem CID 31272
ChEBI CHEBI:31328
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

Absorbance max. 0.01 at 400 nm, max. 0.01 at 320 nm, max. 0.02 at 300 nm, max. 0.04 at 275 nm, max. 0.20 at 260 nm
CAS Max % 100.0000%
Melting Point -78°C (literature)
Density 0.88 g/mL at 25 °C(lit.)
Boiling Point 126.2°C
Flash Point 27°C
Packaging Glass Bottle
Linear Formula CH3COO(CH2)3CH3
Quantity 6 x 1 L
UN Number UN1123
Beilstein 1741921
Formula Weight 116.16
Percent Purity >99.65%
Grade HPLC
Chemical Name or Material Butyl acetate, CHROMASOLV™ Plus, for HPLC, 99.7%
Show More Show Less
ShelfLife 900 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Butyl acetates
EINECSNumber 204-658-1
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.