Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl Acetate, puriss. p.a., ACS Reagent, ≥99.5% (GC), Solstice

Catalog No. 60046924 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-924 1 L Glass Bottle
60-046-925 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-924 Supplier Solstice Supplier No. 458601L
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009445
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
PubChem CID 31272
ChEBI CHEBI:31328
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

Melting Point -78°C
Density 0.88g/mL at 25°C
Boiling Point 126°C
Explosion Limit 0.076
Flash Point 23°C (73.4°F)
Ignition Point 790°F
Packaging Glass Bottle
Refractive Index n20/D 1.394
Linear Formula CH3COO(CH2)3CH3
Quantity 1 L
UN Number UN1123
Residue after Evaporation 0 to 0.001%
Beilstein 1741921
Vapor Density 4 (vs air)
Vapor Pressure 15mmHg (25°C), 8mmHg (20°C)
Formula Weight 116.16g/mol
Percent Purity ≥99.5% (GC)
Chemical Name or Material Butyl acetate
Show More Show Less
Hazard Statement P210-P370 + P378
DOTInformation class 3 - PG 3 - Butyl acetates
RTECSNumber 204-658-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.