Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Butyl acrylate

Catalog No. 2349231L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 L
18 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-492-31L 1 L
23-492-318L 18 L
23-492-3100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-492-31L Supplier Sigma Aldrich Supplier No. 2349231L
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Butyl acrylate is a clear colorless liquid. It is a vinyl monomer. It undergoes radical copolymerization with benzoxazine containing a vinyl group to afford copolymers. Heck coupling reactions of aryl bromides with n-butyl acrylate mediated by phosphine-imidazolium salt has been reported. Copolymerization of styrene and n-butyl acrylate by ATRP catalyzed by CuBr/4,4G-di(5-nonyl)-2,2G-bipyridine has been described.Butyl acrylate is used to prepare: Poly(butyl acrylate) particles.Poly(butyl acrylate-b-acrylic acid) block copolymer.Amphiphilic charged diblock copolymers poly(butyl acrylate)-b-poly(acrylic acid).Poly(n-butyl acrylate), via atom transfer radical polymerization (ATRP) of n-butyl acrylate in the presence of CuIBr/4,4G-di(5-nonyl)-2,2G-bipyridine (catalyst).

Specifications

CAS 141-32-2
Density 0.894 g/mL (at 25°C)
Boiling Point 145°C
Molecular Formula C7H12O2
Refractive Index n20/D 1.418 (literature)
Linear Formula CH2=CHCOO(CH2)3 CH3
Quantity 1 L
MDL Number MFCD00009446
Molecular Weight (g/mol) 128.17
Percent Purity ≥99%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.