Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl Alcohol ACS, MilliporeSigma™

Catalog No. MBX17805 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MBX17805 4 L Glass Bottle
MBX17806 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MBX17805 Supplier MilliporeSigma Supplier No. BX17805
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Quantity 4 L
Packaging Glass Bottle
Linear Formula CH3(CH2)2CH2OH
Formula Weight 74.12
Grade ACS
Chemical Name or Material Butyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.