Learn More
Medchemexpress LLC Butyl benzoate | 136-60-7 | MFCD00009439 | 99.7% | 178.23 g·mol^-1 | C11H14O2 | 250 G
SDP
Catalog No. 5000302699
Encompass_Preferred
Butyl benzoate (n-Butyl Benzoate) is a high-purity aromatic ester (CAS 136-60-7) used as a fragrance ingredient, solvent, and reagent for research and formulation. The product is supplied with a certificate of analysis and safety data sheet, and is available in multiple pack sizes for laboratory use.
  • High purity: 99.73%.
  • Pleasant aromatic odor useful in fragrances and flavor formulations.
  • Acts as an effective organic solvent for cellulose ethers and resins.
  • Supplied with COA and SDS in multiple languages.
  • Available in convenient pack sizes for research (250 g, 500 g, 1 kg).
  • Liquid at room temperature and miscible with common organic solvents.
Catalog No. 50-003-02699 Supplier Medchemexpress LLC Supplier No. 250GHYW016599
Butyl benzoate (n-Butyl Benzoate) is a high-purity aromatic ester (CAS 136-60-7) used as a fragrance ingredient, solvent, and reagent for research and formulation. The product is supplied with a certificate of analysis and safety data sheet, and is available in multiple pack sizes for laboratory use.
  • High purity: 99.73%.
  • Pleasant aromatic odor useful in fragrances and flavor formulations.
  • Acts as an effective organic solvent for cellulose ethers and resins.
  • Supplied with COA and SDS in multiple languages.
  • Available in convenient pack sizes for research (250 g, 500 g, 1 kg).
  • Liquid at room temperature and miscible with common organic solvents.