Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Butyl butyrate

Catalog No. 2819641L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-196-41L 1 L
28-196-4250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 28-196-41L Supplier Sigma Aldrich Supplier No. 2819641L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Butyl butyrate is an ester present in pineapple flavor and is very important for the food and beverages industries
  • Kinetcis of butyl butyrate synthesis by enzymatic esterification and trans-esterification in supercritical carbon dioxide with a lipase, novozym 435 has been reported

Specifications

CAS 109-21-7
Density 0.869 g/mL (at 25°C (literature))
Boiling Point 164°C to 165°C (lit.)
Molecular Formula C8H16O2
Linear Formula CH3CH2CH2COO(CH2)3 CH3
Refractive Index n20/D 1.406 (literature)
MDL Number MFCD00009450
Quantity 1 L
Molecular Weight (g/mol) 144.21
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.