Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl Carbamate 98.0+%, TCI America™
SDP

Catalog No. C0613500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0613500G 500 g
C061325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0613500G Supplier TCI America Supplier No. C0613500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 592-35-8
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.148
MDL Number MFCD00007967
InChI Key SKKTUOZKZKCGTB-UHFFFAOYSA-N
Synonym n-butyl carbamate, carbamic acid, butyl ester, usaf fo-1, usaf el-101, carbamic acid butyl ester, unii-my8i82ajs5, ccris 5069, my8i82ajs5, butyl aminooate, o-butyl carbamate
PubChem CID 11596
IUPAC Name butyl carbamate
SMILES CCCCOC(=O)N

Specifications

Melting Point 54°C
Boiling Point 204°C
Color White
Quantity 500 g
Formula Weight 117.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Butyl Carbamate
EINECSNumber (2)-1712
RTECSNumber EZ0175000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.