Learn More
MSC BUTYL CLEANER/DEGREASER
Catalog No. 502819561
Encompass
This cleaner and degreaser is a liquid, butyl-based concentrate designed for effective cleaning of washable surfaces.
  • Butyl-based formula
  • Dilutable concentrate
  • Suitable for washable surfaces
  • Non-acidic
  • Biodegradable
  • Contains no scent
  • Low VOC content (53 g/L or 5%)
  • Supplied in a 5 gal drum
Catalog No. 50-281-9561 Supplier MSC Supplier No. 33701277
This cleaner and degreaser is a liquid, butyl-based concentrate designed for effective cleaning of washable surfaces.
  • Butyl-based formula
  • Dilutable concentrate
  • Suitable for washable surfaces
  • Non-acidic
  • Biodegradable
  • Contains no scent
  • Low VOC content (53 g/L or 5%)
  • Supplied in a 5 gal drum