Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl Decanoate 98.0+%, TCI America™
SDP

Catalog No. D002025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D002025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D002025ML Supplier TCI America Supplier No. D002025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 30673-36-0
Molecular Formula C14H28O2
Molecular Weight (g/mol) 228.376
MDL Number MFCD00053823
InChI Key ZRNCNTSXSYXHOW-UHFFFAOYSA-N
Synonym Decanoic Acid Butyl Ester
PubChem CID 35408
IUPAC Name butyl decanoate
SMILES CCCCCCCCCC(=O)OCCCC

Specifications

Color Colorless
Boiling Point 123°C
Quantity 25 mL
Formula Weight 228.38
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Decanoate
EINECSNumber (2)-0782
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.