Learn More
Medchemexpress LLC Butyl diphenyl phosphate | 2752-95-6 | MFCD00411621 | >95.0% | 306.29 g/mol | C16H19O4P | 25 MG
SDP
Catalog No. 5000283902
Encompass_Preferred
Butyl diphenyl phosphate is an organophosphate compound used primarily as a flame retardant and plastic additive for research and industrial testing. It is supplied as a small-quantity research reagent with reported analytical-grade purity and is commonly referenced by CAS 2752-95-6 and a molar mass of 306.29 g/mol.
  • Used as a flame retardant and plastic additive.
  • Organophosphate ester chemical structure.
  • High purity suitable for analytical applications (>95%).
  • Available in small research quantities (e.g., 25 MG).
  • Identifiable by CAS number 2752-95-6 and molecular weight 306.29 g/mol.
Catalog No. 50-002-83902 Supplier Medchemexpress LLC Supplier No. HYW54922725MG
Butyl diphenyl phosphate is an organophosphate compound used primarily as a flame retardant and plastic additive for research and industrial testing. It is supplied as a small-quantity research reagent with reported analytical-grade purity and is commonly referenced by CAS 2752-95-6 and a molar mass of 306.29 g/mol.
  • Used as a flame retardant and plastic additive.
  • Organophosphate ester chemical structure.
  • High purity suitable for analytical applications (>95%).
  • Available in small research quantities (e.g., 25 MG).
  • Identifiable by CAS number 2752-95-6 and molecular weight 306.29 g/mol.