Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl Fluoroacetate 97.0+%, TCI America™
SDP

Catalog No. F021525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F021525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F021525ML Supplier TCI America Supplier No. F021525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 371-49-3
Molecular Formula C6H11FO2
Molecular Weight (g/mol) 134.15
MDL Number MFCD00042194
InChI Key FZXXGBGMLALCIH-UHFFFAOYSA-N
Synonym Fluoroacetic Acid Butyl Ester
PubChem CID 67790
IUPAC Name butyl 2-fluoroacetate
SMILES CCCCOC(=O)CF

Specifications

Color Colorless
Boiling Point 154°C
Quantity 25 mL
UN Number 2929
Formula Weight 134.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Fluoroacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.