Learn More
ILC Dover Butyl Hood Assembly for use with Sentinel XL Blower
Catalog No. 19631063
Change view
Click to view available options
For Use With: Sentinel XL Blower
Catalog No.
19-631-063 For Use With: Sentinel XL Blower
1 options
Catalog No. 19-631-063 Supplier ILC Dover Supplier No. S2001
Add to Cart
Add to Cart
Loose-fitting powered air-purifying respirator (PAPR) hood designed to deliver a CAP 2 protection rating when used with the Sentinel XL Blower.
  • Designed to deliver a CAP 2 protection rating when used with the Sentinel XL Blower
Specifications
Product Type Hood Assembly
For Use With (Equipment) Sentinel XL Blower
Model S-2001