Learn More
Medchemexpress LLC Butyl isobutyl phthalate | 17851-53-5 | 100 MG
SDP
Catalog No. 5000417189
Encompass_Preferred
Butyl isobutyl phthalate is isolated from the rhizoid of Laminaria japonica. It is a non-competitive α-glucosidase inhibitor with an IC50 value of 38 μM, showing a hypoglycemic effect and potential for diabetes treatment.
  • Isolated from the rhizoid of Laminaria japonica
  • Non-competitive α-glucosidase inhibitor
  • IC50 value of 38 μM for α-glucosidase
  • Shows a hypoglycemic effect
  • Has potential for diabetes treatment
Catalog No. 50-004-17189 Supplier Medchemexpress LLC Supplier No. HYN7377100MG
Butyl isobutyl phthalate is isolated from the rhizoid of Laminaria japonica. It is a non-competitive α-glucosidase inhibitor with an IC50 value of 38 μM, showing a hypoglycemic effect and potential for diabetes treatment.
  • Isolated from the rhizoid of Laminaria japonica
  • Non-competitive α-glucosidase inhibitor
  • IC50 value of 38 μM for α-glucosidase
  • Shows a hypoglycemic effect
  • Has potential for diabetes treatment