Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl Isobutyrate 98.0+%, TCI America™
SDP

Catalog No. I0315500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0315500ML 500 mL
I031525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0315500ML Supplier TCI America Supplier No. I0315500ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 97-87-0
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00048773
InChI Key JSLCOZYBKYHZNL-UHFFFAOYSA-N
Synonym Isobutyric Acid Butyl Ester
PubChem CID 7353
IUPAC Name butyl 2-methylpropanoate
SMILES CCCCOC(=O)C(C)C

Specifications

Color Colorless
Boiling Point 150°C
Quantity 500 mL
UN Number 3272
Formula Weight 144.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Isobutyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.