Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Butyl isocyanate

Catalog No. B957365ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B957365ML 5 mL
B9573625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B957365ML Supplier Sigma Aldrich Supplier No. B957365ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Butyl isocyanate is a metabolite of benomyl, a broad spectrum fungicide.Butyl isocyanate can undergo anionic polymerization to form poly(butyl isocyanate) using lanthanum isopropoxide as the initiator.

Specifications

CAS 111-36-4
Density 0.88 g/mL (at 25°C (literature))
Molecular Formula C5H9NO
Linear Formula CH3(CH2)3 NCO
MDL Number MFCD00002046
Boiling Point 115°C (lit.)
Synonym 1-Isocyanatobutane; n-Butyl isocyanate; BIC
Molecular Weight (g/mol) 99.13
Quantity 5 mL
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.